accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6284-40-8

H3CNHOHOHOHOHOH

CAS RN 6284-40-8

IUPAC Name: 6-(methylamino)hexane-1,2,3,4,5-pentol
Synonyms: methylglucamine
Molecular Weight (g/mol): 195.22
Molecular Formula: C7H17NO5
InChi Key: MBBZMMPHUWSWHV-UHFFFAOYNA-N
SMILES: CNCC(O)C(O)C(O)C(O)CO
Molecular Weight (g/mol) 
  • (6)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (6)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)