accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 628690-75-5

CH3CH3NNFFNFHO

CAS RN 628690-75-5

IUPAC Name: 2',4-difluoro-5'-[8-fluoro-7-(2-hydroxypropan-2-yl)imidazo[1,2-a]pyridin-3-yl]-[1,1'-biphenyl]-2-carbonitrile
Synonyms: 2',4-difluoro-5'-[8-fluoro-7-(2-hydroxypropan-2-yl)imidazo[1,2-a]pyridin-3-yl]-[1,1'-biphenyl]-2-carbonitrile
Molecular Weight (g/mol): 407.4
Molecular Formula: C23H16F3N3O
InChi Key: SJMMDWXJSSJHOQ-UHFFFAOYSA-N
SMILES: CC(C)(O)C1=C(F)C2=NC=C(N2C=C1)C1=CC(=C(F)C=C1)C1=C(C=C(F)C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results