accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 629-04-9

H3CBr

CAS RN 629-04-9

IUPAC Name: 1-bromoheptane
Synonyms: 1-bromoheptane
Molecular Weight (g/mol): 179.1
Molecular Formula: C7H15Br
InChi Key: LSXKDWGTSHCFPP-UHFFFAOYSA-N
SMILES: CCCCCCCBr
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)