accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 629-12-9

IUPAC Name: 1-isothiocyanatopentane
Synonyms: 1-isothiocyanatopentane
Molecular Weight (g/mol): 129.22
Molecular Formula: C6H11NS
InChi Key: SGHJUJBYMSVAJY-UHFFFAOYSA-N
SMILES: CCCCCN=C=S
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)