accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH2

CAS RN 629-73-2

IUPAC Name: hexadec-1-ene
Synonyms: hexadecene
Molecular Weight (g/mol): 224.43
Molecular Formula: C16H32
InChi Key: GQEZCXVZFLOKMC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCC=C
Molecular Weight (g/mol) 
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results