accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 629-74-3

IUPAC Name: hexadec-1-yne
Synonyms: 1-hexadecyne
Molecular Weight (g/mol): 222.42
Molecular Formula: C16H30
InChi Key: UCIDYSLOTJMRAM-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCC#C
Boiling Point 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)