accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 629-78-7

IUPAC Name: heptadecane
Synonyms: heptadecane N-heptadecane heptadekan
Molecular Weight (g/mol): 240.48
Molecular Formula: C17H36
InChi Key: NDJKXXJCMXVBJW-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)