accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6291-84-5

H3CHNNH2

CAS RN 6291-84-5

IUPAC Name: (3-aminopropyl)(methyl)amine
Synonyms: 1,3-propanediamine, N-methyl-
Molecular Weight (g/mol): 88.15
Molecular Formula: C4H12N2
InChi Key: QHJABUZHRJTCAR-UHFFFAOYSA-N
SMILES: CNCCCN
Boiling Point 
  • (5)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)