accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSCH3

CAS RN 6294-31-1

IUPAC Name: 1-(hexylsulfanyl)hexane
Synonyms: hexane, 1,1'-thiobis-
Molecular Weight (g/mol): 202.4
Molecular Formula: C12H26S
InChi Key: LHNRHYOMDUJLLM-UHFFFAOYSA-N
SMILES: CCCCCCSCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results