accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6294-89-9

IUPAC Name: methoxycarbohydrazide
Synonyms: hydrazinecarboxylic acid, methyl ester
Molecular Weight (g/mol): 90.08
Molecular Formula: C2H6N2O2
InChi Key: WFJRIDQGVSJLLH-UHFFFAOYSA-N
SMILES: COC(=O)NN
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)