CAS RN 62948-75-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Uric Acid-1,3-15N2, TRC
CAS: 62948-75-8 Molecular Formula: C5 H4 15N2 N2 O3 Molecular Weight (g/mol): 170.1 Synonym: 7,9-dihydro-1H-Purine-2,6,8(3H)-trione-1,3-15N2 IUPAC Name: 7,9-dihydro-3H-purine-2,6,8-trione SMILES: O=C1[15NH]C(=O)C2=C([15NH]1)NC(=O)N2