accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 62965-10-0

H3CH3CCH3(S)OHOHNOO

CAS RN 62965-10-0

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-3,3-dimethylbutanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-3,3-dimethylbutanoic acid
Molecular Weight (g/mol): 265.31
Molecular Formula: C14H19NO4
InChi Key: NSVNKQLSGGKNKB-UHFFFAOYNA-N
SMILES: CC(C)(C)C(NC(=O)OCC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)