CAS RN 6299-67-8
IUPAC Name:
2,3-dimethoxyaniline
Synonyms:
2,3-dimethoxyaniline
Molecular Weight (g/mol):
153.18
Molecular Formula:
C8H11NO2
InChi Key:
HEZIOZBMPKPOER-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(N)=C1OC
1
–
1
of
1
results
1
2,3-Dimethoxyaniline, TRC
CAS: 6299-67-8 Molecular Formula: C8H11NO2 Molecular Weight (g/mol): 153.18 Synonym: NSC 44873,[2,3-Bis(methyloxy)phenyl]amine,2,3-Dimethoxybenzenamine IUPAC Name: 2,3-dimethoxyaniline SMILES: O(C1=CC=CC(N)=C1OC)C