accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNH(R)(R)O(S)H3C(R)OCH3NHNON

CAS RN 62996-74-1

IUPAC Name: (2S,3R,4R,6R)-3-methoxy-2-methyl-4-(methylamino)-29-oxa-1,7,17-triazaoctacyclo[12.12.2.1²,⁶.0⁷,²⁸.0⁸,¹³.0¹⁵,¹⁹.0²⁰,²⁷.0²¹,²⁶]nonacosa-8,10,12,14(28),15(19),20(27),21(26),22,24-nonaen-16-one
Synonyms: staurosporine staurosporin (+)-staurosporine
Molecular Weight (g/mol): 466.54
Molecular Formula: C28H26N4O3
InChi Key: HKSZLNNOFSGOKW-FYTWVXJKSA-N
SMILES: CN[C@@H]1C[C@H]2O[C@@](C)([C@@H]1OC)N1C3=C(C=CC=C3)C3=C1C1=C(C4=CC=CC=C4N21)C1=C3CNC1=O
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (7)
Percent Purity 
  • (1)
  • (3)
  • (3)
  • (2)
Physical Form 
  • (4)
  • (3)
Color 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (7)
  • (1)
  • (3)
  • (3)
  • (2)
  • (4)
  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)