accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 63-37-6

IUPAC Name: {[5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid
Synonyms: poly C monophosphate, cytidine cmp (cytidine monophosphate) cytidylic acids cytidylic acid (substance) cytidylate acid, cytidylic acids, cytidylic cmp (nucleotide) cytidine 5'-phosphoric acid
Molecular Weight (g/mol): 323.2
Molecular Formula: C9H14N3O8P
InChi Key: IERHLVCPSMICTF-UHFFFAOYNA-N
SMILES: NC1=NC(=O)N(C=C1)C1OC(COP(O)(O)=O)C(O)C1O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results