accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CS(S)NH2OHO

CAS RN 63-68-3

IUPAC Name: 2-amino-4-(methylsulfanyl)butanoic acid
Synonyms: methionine methionin metionina Hmet
Molecular Weight (g/mol): 149.21
Molecular Formula: C5H11NO2S
InChi Key: FFEARJCKVFRZRR-UHFFFAOYNA-N
SMILES: CSCCC(N)C(O)=O
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (8)
  • (1)
  • (3)
  • (1)
  • (1)
  • (5)
Percent Purity 
  • (7)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (8)
  • (4)
  • (6)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (8)
  • (1)
  • (3)
  • (1)
  • (1)
  • (5)
  • (7)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)