accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3H3CBr

CAS RN 630-17-1

IUPAC Name: 1-bromo-2,2-dimethylpropane
Synonyms: propane, 1-bromo-2,2-dimethyl-
Molecular Weight (g/mol): 151.05
Molecular Formula: C5H11Br
InChi Key: CQWYAXCOVZKLHY-UHFFFAOYSA-N
SMILES: CC(C)(C)CBr
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)