accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3CH3N

CAS RN 630-18-2

IUPAC Name: 2,2-dimethylpropanenitrile
Synonyms: pivalonitrile
Molecular Weight (g/mol): 83.13
Molecular Formula: C5H9N
InChi Key: JAMNHZBIQDNHMM-UHFFFAOYSA-N
SMILES: CC(C)(C)C#N
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)