accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3OCH3CH3OCH3CH3OCH3CH3OCe

CAS RN 63007-83-0

IUPAC Name: λ⁴-cerium(4+) tetrakis(propan-2-olate)
Synonyms: λ⁴-cerium(4+) tetrakis(propan-2-olate)
Molecular Weight (g/mol): 376.47
Molecular Formula: C12H28CeO4
InChi Key: BCBBBOXRROHVFG-UHFFFAOYSA-N
SMILES: [Ce+4].CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results