accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 63010-72-0

FClN

CAS RN 63010-72-0

IUPAC Name: 4-chloro-8-fluoroquinoline
Synonyms: 4-chloro-8-fluoroquinoline
Molecular Weight (g/mol): 181.59
Molecular Formula: C9H5ClFN
InChi Key: NUIMKHDPMIPYFI-UHFFFAOYSA-N
SMILES: FC1=C2N=CC=C(Cl)C2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results