accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOOBrFFF

CAS RN 630125-49-4

IUPAC Name: 1-bromo-3-nitro-5-(trifluoromethyl)benzene
Synonyms: 1-bromo-3-nitro-5-(trifluoromethyl)benzene
Molecular Weight (g/mol): 270.01
Molecular Formula: C7H3BrF3NO2
InChi Key: YHTVYRKVFAFVLP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Br)=CC(=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results