accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 63039-46-3

CH3H3CCH3OOHN(R)CHOO

CAS RN 63039-46-3

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}pent-4-ynoate
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]pent-4-ynoate
Molecular Weight (g/mol): 212.23
Molecular Formula: C10H14NO4
InChi Key: AMKHAJIFPHJYMH-SSDOTTSWSA-M
SMILES: CC(C)(C)OC(=O)N[C@H](CC#C)C([O-])=O
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)