CAS RN 63056-20-2
IUPAC Name:
6-phenylpyridine-3-carbaldehyde
Synonyms:
6-phenylpyridine-3-carbaldehyde
Molecular Weight (g/mol):
183.21
Molecular Formula:
C12H9NO
InChi Key:
HWDMDOSKSDLXEH-UHFFFAOYSA-N
SMILES:
O=CC1=CC=C(N=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
6-Phenylnicotinaldehyde, 97%, Thermo Scientific™
CAS: 63056-20-2 Molecular Formula: C12H9NO Molecular Weight (g/mol): 183.21 MDL Number: MFCD03086130 InChI Key: HWDMDOSKSDLXEH-UHFFFAOYSA-N Synonym: 6-phenylnicotinaldehyde,6-phenylpyridine-3-carboxaldehyde,2-phenyl-5-pyridinecarboxaldehyde,6-phenyl-3-pyridinecarboxaldehyde,3-pyridinecarboxaldehyde, 6-phenyl,2-phenylpyridine-5-carbaldehyde,6-phenyl-pyridine-3-carbaldehyde PubChem CID: 2764562 IUPAC Name: 6-phenylpyridine-3-carbaldehyde SMILES: C1=CC=C(C=C1)C2=NC=C(C=C2)C=O