accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C(S)OCH3NH2CH3OHCl

CAS RN 6306-52-1

IUPAC Name: methyl 2-amino-3-methylbutanoate hydrochloridyl
Synonyms: methyl 2-amino-3-methylbutanoate hydrochloridyl
Molecular Weight (g/mol): 166.63
Molecular Formula: C6H13ClNO2
InChi Key: RFMIEHOYULVMNZ-UHFFFAOYNA-N
SMILES: [Cl].COC(=O)C(N)C(C)C
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (4)
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)