accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClNCl

CAS RN 6306-60-1

IUPAC Name: 2-(2,4-dichlorophenyl)acetonitrile
Synonyms: 2-(2,4-dichlorophenyl)acetonitrile
Molecular Weight (g/mol): 186.04
Molecular Formula: C8H5Cl2N
InChi Key: VJARIBGMDPJLCL-UHFFFAOYSA-N
SMILES: ClC1=CC=C(CC#N)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results