accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 63069-50-1

NH2FN

CAS RN 63069-50-1

IUPAC Name: 4-amino-3-fluorobenzonitrile
Synonyms: 4-amino-3-fluorobenzonitrile
Molecular Weight (g/mol): 136.13
Molecular Formula: C7H5FN2
InChi Key: RLMBRRQWBTWGMB-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1F)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results