accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOS

CAS RN 6314-28-9

IUPAC Name: 1-benzothiophene-2-carboxylic acid
Synonyms: 1-benzothiophene-2-carboxylic acid
Molecular Weight (g/mol): 178.21
Molecular Formula: C9H6O2S
InChi Key: DYSJMQABFPKAQM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=CC=CC=C2S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results