accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6315-89-5

IUPAC Name: 3,4-dimethoxyaniline
Synonyms: benzenamine, 3,4-dimethoxy-
Molecular Weight (g/mol): 153.18
Molecular Formula: C8H11NO2
InChi Key: LGDHZCLREKIGKJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(N)C=C1OC
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)