accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OONHONOOOH

CAS RN 6317-97-1

IUPAC Name: ethyl 6-hydroxy-5-nitro-4-oxo-1,4-dihydropyridine-3-carboxylate
Synonyms: ethyl 6-hydroxy-5-nitro-4-oxo-1H-pyridine-3-carboxylate
Molecular Weight (g/mol): 228.16
Molecular Formula: C8H8N2O6
InChi Key: JBTWWCUXSHUDEJ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CNC(O)=C(C1=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results