accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3HNONCH3H3C

CAS RN 632-14-4

IUPAC Name: 1,3,3-trimethylurea
Synonyms: urea, trimethyl-
Molecular Weight (g/mol): 102.14
Molecular Formula: C4H10N2O
InChi Key: COSWCAGTKRUTQV-UHFFFAOYSA-N
SMILES: CNC(=O)N(C)C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)