accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 632-16-6

CH3SH3C

CAS RN 632-16-6

IUPAC Name: 2,3-dimethylthiophene
Synonyms: thiophene, 2,3-dimethyl-
Molecular Weight (g/mol): 112.19
Molecular Formula: C6H8S
InChi Key: BZYUMXXOAYSFOW-UHFFFAOYSA-N
SMILES: CC1=C(C)C=CS1
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)