accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 63214-57-3

NHONO

CAS RN 63214-57-3

IUPAC Name: 4-(piperidine-4-carbonyl)morpholine
Synonyms: 4-(piperidine-4-carbonyl)morpholine
Molecular Weight (g/mol): 198.27
Molecular Formula: C10H18N2O2
InChi Key: UVDVXQHUTBCRKT-UHFFFAOYSA-N
SMILES: O=C(C1CCNCC1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results