accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6324-11-4

IUPAC Name: 2-(2-hydroxyphenoxy)acetic acid
Synonyms: O-hydroxyphenoxyacetic acid
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: LPXHPAYRONCQIF-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC=CC=C1O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results