accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SOONOO

CAS RN 6325-93-5

IUPAC Name: 4-nitrobenzene-1-sulfonamide
Synonyms: 4-nitrobenzenesulfonamide
Molecular Weight (g/mol): 202.18
Molecular Formula: C6H6N2O4S
InChi Key: QWKKYJLAUWFPDB-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results