accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 633-03-4

CH3NCH3CNCH3H3COHSOOO

CAS RN 633-03-4

IUPAC Name: bis[4-(diethylamino)phenyl](phenyl)methylium hydrogen sulfate
Synonyms: brilliant green (dye)
Molecular Weight (g/mol): 482.64
Molecular Formula: C27H34N2O4S
InChi Key: NNBFNNNWANBMTI-UHFFFAOYSA-M
SMILES: OS([O-])(=O)=O.CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(CC)CC
Quantity 
  • (3)
  • (3)
  • (1)
Color 
  • (4)
Grade 
  • (1)
  • (3)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)