accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6331-04-0

CH3OHOH3C

CAS RN 6331-04-0

IUPAC Name: 2-(2,4-dimethylphenyl)acetic acid
Synonyms: (2,4-dimethylphenyl)acetic acid
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: MWBXCWLRASBZFB-UHFFFAOYSA-N
SMILES: CC1=CC=C(CC(O)=O)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results