accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH3CONH2

CAS RN 6331-71-1

IUPAC Name: 4-amino-N,N-dimethylbenzamide
Synonyms: 4-amino-N,N-dimethylbenzamide
Molecular Weight (g/mol): 164.21
Molecular Formula: C9H12N2O
InChi Key: QEPGWLBMAAEBCP-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results