accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6336-68-1

IUPAC Name: 4-(piperidine-1-sulfonyl)aniline
Synonyms: 4-(piperidine-1-sulfonyl)aniline
Molecular Weight (g/mol): 240.32
Molecular Formula: C11H16N2O2S
InChi Key: ZTTBIWZAAMPNBE-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)S(=O)(=O)N1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results