CAS RN 63411-78-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(4-Pyridyl)-7-azabenzimidazole, 97%, Thermo Scientific™
CAS: 63411-78-9 Molecular Formula: C11H8N4 Molecular Weight (g/mol): 196.213 MDL Number: MFCD01001152 InChI Key: NEAHTABRXFKZGG-UHFFFAOYSA-N Synonym: 4-1h-imidazo 4,5-c pyridin-2-yl pyridine,cambridge id 5237049,2-4-pyridyl-7-azabenzimidazole,2-4-pyridyl-3h-imidazo 4,5-c pyridine,2-pyridin-4-yl-3h-imidazo 4,5-c pyridine,2-4-pyridinyl-1h-imidazo 4,5-c pyridine,4-3h-imidazo 4,5-c pyridin-2-yl pyridine,2-pyridin-4-yl-1h-imidazo 4,5-c pyridine,3h-imidazo 4,5-c pyridine,2-4-pyridinyl PubChem CID: 789950 IUPAC Name: 2-pyridin-4-yl-3H-imidazo[4,5-c]pyridine SMILES: C1=CN=CC=C1C2=NC3=C(N2)C=NC=C3