accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6342-21-8

IUPAC Name: (2-amino-1-phenylethyl)dimethylamine
Synonyms: (2-amino-1-phenylethyl)dimethylamine
Molecular Weight (g/mol): 164.25
Molecular Formula: C10H16N2
InChi Key: NFSAPTWLWWYADB-UHFFFAOYNA-N
SMILES: CN(C)C(CN)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results