accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 63425-54-7

IUPAC Name: 3-chloro-1-methyl-1H-pyrazole
Synonyms: 3-chloro-1-methylpyrazole
Molecular Weight (g/mol): 116.55
Molecular Formula: C4H5ClN2
InChi Key: GWZMWAZUCSGVLQ-UHFFFAOYSA-N
SMILES: CN1C=CC(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results