accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 63449-41-2

IUPAC Name: benzyl(decyl)dimethylazanium chloride
Synonyms: benzyl(decyl)dimethylazanium chloride
Molecular Weight (g/mol): 311.94
Molecular Formula: C19H34ClN
InChi Key: TTZLKXKJIMOHHG-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results