accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NH2NHONH2NH2NHOOHSOHOO

CAS RN 6345-29-5

IUPAC Name: bis(N''-hydroxyguanidine); sulfuric acid
Synonyms: bis(2-hydroxyguanidine); sulfuric acid
Molecular Weight (g/mol): 248.21
Molecular Formula: C2H12N6O6S
InChi Key: MTGDDPZRXSDPFH-UHFFFAOYSA-N
SMILES: NC(N)=NO.NC(N)=NO.OS(O)(=O)=O
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results