CAS RN 63451-35-4
IUPAC Name:
trisodium 3-hydroxy-4-{2-[(1Z)-2-oxo-4-sulfonato-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}naphthalene-2,7-disulfonate
Synonyms:
hydroxynaphthol blue
Molecular Weight (g/mol):
620.46
Molecular Formula:
C20H11N2Na3O11S3
InChi Key:
JQECMNGWLIFQLL-QRQZZYIYSA-K
SMILES:
[Na+].[Na+].[Na+].OC1=C(C=C2C=C(C=CC2=C1N\N=C1/C(=O)C=C(C2=CC=CC=C12)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O
1
–
2
of
2
results
1
Promotions Available
Hydroxynaphthol Blue, Indicator for Metal Titration, Solstice
Hydroxynaphthol Blue, Indicator for Metal Titration, Solstice
CAS: 63451-35-4 Molecular Formula: C20H11N2Na3O11S3 Molecular Weight (g/mol): 620.46 MDL Number: MFCD00036393 InChI Key: JQECMNGWLIFQLL-UHFFFAOYSA-K PubChem CID: 44134769 IUPAC Name: trisodium;3-hydroxy-4-[2-(2-oxo-4-sulfonatonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonate SMILES: C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+]
2
Hydroxynapthol Blue (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.