CAS RN 635-78-9
IUPAC Name:
7-hydroxy-3H-phenoxazin-3-one
Synonyms:
resorufin resorufine
Molecular Weight (g/mol):
213.19
Molecular Formula:
C12H7NO3
InChi Key:
HSSLDCABUXLXKM-UHFFFAOYSA-N
SMILES:
OC1=CC=C2N=C3C=CC(=O)C=C3OC2=C1
1
–
1
of
1
results
1
Resorufin, TRC
CAS: 635-78-9 Molecular Formula: C12H7NO3 Molecular Weight (g/mol): 213.19 Synonym: 7-Hydroxy-3H-phenoxazin-3-one,7-Hydroxyphenoxaz-3-one,7-Hydroxyphenoxazin-3-one,NSC 12097,Resorufin,NSC 12097,Resofurin,Resorufine IUPAC Name: 7-hydroxy-3H-phenoxazin-3-one SMILES: O=C(C=C1)C=C2C1=NC3=CC=C(O)C=C3O2