CAS RN 6351-10-6
IUPAC Name:
2,3-dihydro-1H-inden-1-ol
Synonyms:
indanol (+-)-1-indanol 1-indanol, (+-)- (s)-1-indanol (S)-1-indanol (r)-1-indanol (R)-1-indanol 1-indanol, (S)- 1-indanol, (s)- 1-indanol, (R)-
Molecular Weight (g/mol):
134.18
Molecular Formula:
C9H10O
InChi Key:
YIAPLDFPUUJILH-UHFFFAOYNA-N
SMILES:
OC1CCC2=CC=CC=C12
1
–
1
of
1
results
1
1-Indanol, TRC
CAS: 6351-10-6 Molecular Formula: C9 H10 O Molecular Weight (g/mol): 134.175 Synonym: 1H-Inden-1-ol, 2,3-dihydro-,1-Indanol (6CI,7CI,8CI),1-Indanol, (±)-,2,3-Dihydro-1H-inden-1-ol,(RS)-1-Indanol,(±)-1-Indanol,(±)-2,3-Dihydro-1H-inden-1-ol,1-Hydroxyhydrindene,1-Hydroxyindan,NSC 31258 IUPAC Name: 2,3-dihydro-1H-inden-1-ol SMILES: OC1CCc2ccccc12