accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaCH3ONOHN(S)(S)SNOOOOCH3OSH2NN(Z)

CAS RN 63527-52-6

IUPAC Name: sodium (6S,7S)-3-[(acetyloxy)methyl]-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonyms: cefotaxime
Molecular Weight (g/mol): 477.44
Molecular Formula: C16H16N5NaO7S2
InChi Key: AZZMGZXNTDTSME-XJTDMATHSA-M
SMILES: [Na+].CO\N=C(/C(=O)N[C@@H]1[C@@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results