accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaNaNaHOSOOOSOOOSOOO

CAS RN 6358-69-6

IUPAC Name: trisodium 8-hydroxypyrene-1,3,6-trisulfonate
Synonyms: pyranine solvent green 7 japan green 204
Molecular Weight (g/mol): 524.37
Molecular Formula: C16H7Na3O10S3
InChi Key: KXXXUIKPSVVSAW-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].OC1=CC(=C2C=CC3=C(C=C(C4=CC=C1C2=C34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (5)
  • (5)
Quantity 
  • (2)
  • (1)
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (3)
Grade 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (5)
  • (5)
  • (2)
  • (1)
  • (3)
  • (4)
  • (2)
  • (3)
  • (3)
  • (3)