accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 636-97-5

IUPAC Name: 4-nitrobenzohydrazide
Synonyms: 4-nitrobenzoylhydrazine
Molecular Weight (g/mol): 181.15
Molecular Formula: C7H7N3O3
InChi Key: FKZXYJYTUSGIQE-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results