accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CHO

CAS RN 63697-96-1

IUPAC Name: 4-ethynylbenzaldehyde
Synonyms: 4-ethynylbenzaldehyde
Molecular Weight (g/mol): 130.15
Molecular Formula: C9H6O
InChi Key: BGMHQBQFJYJLBP-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)C#C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results